Microplates, copper coated, 96-well, Pierce™
Supplier: Thermo Fisher Scientific
15143 15146 15147 15148
PIER15143EA 375 EUR
PIER15143 PIER15146 PIER15147 PIER15148
Microplates, copper coated, 96-well, Pierce™
Microplates Immunology Microplates and Strips
Pierce Copper coated plates are ideal for analysing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper.
  • Quantitate previously undetectable histidine-tagged proteins
  • Wider dynamic range with four-fold greater capacity than regular nickel chelate-coated plate
  • Copper chelate provides greater binding capacity than nickel
  • Detection limit is 0,1 pg of polyhistidine tagged fusion protein per well
  • Pre-