NHS-Activated magnetic beads, Pierce™
Supplier: Thermo Fisher Scientific
88826 88827
PIER88826EA 344 EUR
PIER88826 PIER88827
NHS-Activated magnetic beads, Pierce™
Beads Magnetic Beads
The Pierce™ NHS-activated magnetic beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use in custom affinity purification experiments.
  • High Capacity—at least four times higher binding capacity than NHS-Activated Magnetic Beads from other suppliers
  • Easy to use—Immobilise in a simple one-step reaction with minimal hands-on time
  • Safe—No hazardous chemicals needed (e.g., Sodium Cyanoborohydride and Cyanogen Bromide)
  • Ligand-Compatible—use to immobilise with nearly any primary amine-containing compound