Peptide desalting spin columns, Pierce™
Supplier: Thermo Fisher Scientific
89851 89852
PIER89851EA 463 EUR
PIER89851 PIER89852
Peptide desalting spin columns, Pierce™
Chromatography Columns
Thermo Scientific™ Pierce™ peptide desalting spin columns are ready to use centrifuge spin columns that enable efficient desalting of peptide samples following enzymatic digestion. The polymer-based hydrophobic resin contained in each column provides excellent binding and recovery characteristics for peptide samples in preparation for mass spectrometry and other methods.
  • Removes MS-interfering contaminants: Significantly reduces signal suppression and improves sensitivity in mass spectrometry
  • High binding capacity: Special hydrophobic polymer-based resin