4'-Ethoxyacetophenone

Supplier: LGC Standards PROMOCHEM

MM0474.04
PROOMM0474.04EA 321 EUR
PROOMM0474.04
4'-Ethoxyacetophenone
4'-Ethoxyacetophenone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 83 °C (0.04 mmHg)
Melting Pt: 39
MDL Number: MFCD00009095
CAS Number: 1676-63-7
EINECS: 216-825-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR