Probenecid

Supplier: LGC Standards PROMOCHEM

Synonyms: 4-[(dipropylamino)sulphonyl]benzoic acid

MM0657.00
PROOMM0657.00EA 1260 EUR
PROOMM0657.00
Probenecid
Probenecid
Formula: C₁₃H₁₉NO₄S
MW: 285.4 g/mol
MDL Number: MFCD00038402
CAS Number: 57-66-9
EINECS: 200-344-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR