Indium standard solution
Supplier: LGC Standards PROMOCHEM
NIST-3124A
PROONIST-3124AEA 1140 EUR
PROONIST-3124A
Indium standard solution
Standards Cationic Standards Indium Standards
This reference standard is intended for use as a primary calibration standard for the quantitative determination of indium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of indium.
Certifikace: Traceable to NIST and SRM numbers.