Organic reference standards, cyclazadone (NMIA D914), Certified Reference Material (CRM)

Supplier: LGC Standards PROMOCHEM

NMIAD914
PROONMIAD914EA 266 EUR
PROONMIAD914
Organic reference standards, cyclazadone (NMIA D914), Certified Reference Material (CRM)
Standards Organic Standards

The purity value obtained from a combination of traditional analytical techniques is 99,6 ± 1,4% mass fraction, (95% coverage interval). The purity estimate by traditional analytical techniques was obtained by subtraction from 100% of total impurities by GC-FID, thermogravimetric analysis, Karl Fischer analysis and 1H NMR and also supported by elemental microanalysis.


  • White solid
  • Moisture content ≤0,3% mass fraction
  • Volatile residue ≤0,1% mass fraction
  • Non volatile residue ≤0,2% mass fraction
  • CAS no.: 14461-91-7


Synonyms: 5-phenyl-2-cyclopropylamino-4-oxazolinone; 4(5H)-oxazolone-2-(cyclopropylamino)-5-phenyl-cyclopropylpemoline.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR