Boron standard solution, for ICP, in H₂O, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PBW-100 VHG-PBW-50 VHG-PBW-500 VHG-LBW-100
PROOVHG-PBW-100EA 53.5 EUR
PROOVHG-PBW-100 PROOVHG-PBW-50 PROOVHG-PBW-500 PROOVHG-LBW-100
Boron standard solution, for ICP, in H₂O, VHG
Standards Cationic Standards Boron Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikace: Traceable to NIST, SRM.