Ruthenium standard solution, for ICP, in 20% HCl, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PRUH-100 VHG-PRUH-500 VHG-TRUH-100 VHG-TRUH-500
PROOVHG-PRUH-100EA 110 EUR
PROOVHG-PRUH-100 PROOVHG-PRUH-500 PROOVHG-TRUH-100 PROOVHG-TRUH-500
Ruthenium standard solution, for ICP, in 20% HCl, VHG
Standards Cationic Standards Ruthenium Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikace: Traceable to NIST, SRM.