Lanthanum standard solution, for ICP, in 5% HNO₃, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PLAN-100 VHG-PLAN-500 VHG-TLAN-100 VHG-TLAN-500
PROOVHG-PLAN-100EA 55.7 EUR
PROOVHG-PLAN-100 PROOVHG-PLAN-500 PROOVHG-TLAN-100 PROOVHG-TLAN-500
Lanthanum standard solution, for ICP, in 5% HNO₃, VHG
Standards Cationic Standards Lanthanum Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikace: Traceable to NIST, SRM.