DX4™ Work Trousers
Supplier: Portwest
DX449BKR28 DX449BKR30 DX449BKR32 DX449BKR33 DX449BKR34 DX449BKR36 DX449BKR38 DX449BKR40 DX449BKR41 DX449BKR42 DX449BKR44 DX449BKR46 DX449BKR48 DX449MBR28 DX449MBR30 DX449MBR32 DX449MBR33 DX449MBR34 DX449MBR36 DX449MBR38 DX449MBR40 DX449MBR41 DX449MBR42 DX449MBR44 DX449MBR46 DX449MBR48
PRTWDX449BKR28EA 110 EUR
PRTWDX449BKR28 PRTWDX449BKR30 PRTWDX449BKR32 PRTWDX449BKR33 PRTWDX449BKR34 PRTWDX449BKR36 PRTWDX449BKR38 PRTWDX449BKR40 PRTWDX449BKR41 PRTWDX449BKR42 PRTWDX449BKR44 PRTWDX449BKR46 PRTWDX449BKR48 PRTWDX449MBR28 PRTWDX449MBR30 PRTWDX449MBR32 PRTWDX449MBR33 PRTWDX449MBR34 PRTWDX449MBR36 PRTWDX449MBR38 PRTWDX449MBR40 PRTWDX449MBR41 PRTWDX449MBR42 PRTWDX449MBR44 PRTWDX449MBR46 PRTWDX449MBR48
DX4™ Work Trousers
Workwear
The DX4 work trouser is ergonomically designed and uses the targeted placement of dynamic 4X stretch fabrics to give maximum range of movement when working. Triple stitching throughout for maximum durability.
  • 4-way stretch fabric for ease of movement and added comfort
  • Lightweight flexible HiVisTex Pro segmented reflective tape for increased visibility
  • 11 pockets for ample storage
  • Rule pocket
  • Pre-bent knees allow for increased freedom of movement
  • Two tier knee pad pockets allowing two positioning options
  • Side elastic waist for ultimate wearer comfort
  • High