Chemical pads
Supplier: Portwest
SM80YER
PRTWSM80YEREA 161 EUR
PRTWSM80YER
Chemical pads
Spill Control Materials
Perforated single spun bonded layers gives excellent tensile strength and rapid absorption properties.
  • Suitable for use in lint free environments
  • Each pad can absorb 0,4 L of aggressive and non-aggressive chemicals
Certifikace: Meets standards BS 7959-1 and BS 7959-3.