Nitrogen flasks, round bottom with NS stopcock
Supplier: QVF LABORTECHNIK
GSG01024H07 GSG01024H08 GSG01020H02 GSG01020H03 GSG01020H06 GSG01020H07
QVFLGSG01024H07EA 0 EUR
QVFLGSG01024H07 QVFLGSG01024H08 QVFLGSG01020H02 QVFLGSG01020H03 QVFLGSG01020H06 QVFLGSG01020H07
Nitrogen flasks, round bottom with NS stopcock
Flasks Nitrogen Flasks
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With conical ground joint, NS stopcock and inscription label.