Stoppers, for test tubes
Supplier: RATIOLAB
3514900 351490B 351490G 351490R 3914900
RATI3514900EA 16.61 EUR
RATI3514900 RATI351490B RATI351490G RATI351490R RATI3914900
Stoppers, for test tubes
Stoppers PE/LDPE/HDPE Stoppers
PE.
  • Very tight seal due to two sealing rings
  • Available in natural and four more colors