Ternary tee mixer
Supplier: Restek
25138
REST25138EA 1079 EUR
REST25138
Ternary tee mixer
Shakers Vortex Shakers
This stainless steel ternary tee mixer is an efficient cross-flow shearing mechanism.
  • Reduced baseline noise
  • Increased sensitivity
  • Increased reaction efficiency in post-column derivatisation
  • Improved gradient accuracy for more reproducible results