606 Phthalate esters calibration mix (6 components)
Supplier: Restek
31031
REST31031EA 65 EUR
REST31031
606 Phthalate esters calibration mix (6 components)
Standards Environmental Analysis Standards
Calibration mix contains Bis(2-ethylhexyl)phthalate, benzyl butyl phthalate, di-n-butyl phthalate, diethylphthalate, dimethylphthalate, di-n-octyl phthalate.
Certifikace: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.