Extraction solvent stock solution #1 (2 components)
Supplier: Restek
31679
REST31679EA 72.5 EUR
REST31679
Extraction solvent stock solution #1 (2 components)
Standards Environmental Analysis Standards
This extraction solvent stock solution contains: n-decane (C10), n-tetracontane (C40).