L/C pesticide lab control sample (7 components)
Supplier: Restek
32040
REST32040EA 67 EUR
REST32040
L/C pesticide lab control sample (7 components)
Standards Organic Standards
This sample contains seven components.
Components are γ-BHC (Lindane) (10 μg/ml), trans-Chlordane (10 μg/ml), 4,4'-DDE (20 μg/ml), dieldrin (20 μg/ml), endosulphan sulphate (20 μg/ml), endrin (20 μg/ml), heptachlor epoxide (isomer B) (10 μg/ml).
Certifikace: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.