UHPLC columns, Pinnacle® DB Cyano (USP L10)
Supplier: Restek
9416512-700 9416513-700 9416515-700 9416522-700 9416523-700 9416525-700 9416532-700 9416533-700 9416535-700 9416552-700 9416553-700 9416555-700 9416562-700 9416563-700 9416565-700 9416572-700 9416573-700 9416575-700 9416510 9416511 9416512 9416513 9416515 9416517 9416518 9416519 9416521 9416522 9416523 9416525 9416531 9416532 9416533 9416535 9416550 9416551 9416552 9416553 9416555 9416557 9416558 9416559 9416560 9416561 9416562 9416563 9416565 9416567 9416568 9416569 9416570 9416571 9416572 9416573 9416575 9416577 9416578 9416579 941650210 941650212 941650220 941650222 9416212 9416232 9416252 941651E 941652E 941653E 941655E 941656E 941657E
REST9416512-700EA 603 EUR
REST9416512-700 REST9416513-700 REST9416515-700 REST9416522-700 REST9416523-700 REST9416525-700 REST9416532-700 REST9416533-700 REST9416535-700 REST9416552-700 REST9416553-700 REST9416555-700 REST9416562-700 REST9416563-700 REST9416565-700 REST9416572-700 REST9416573-700 REST9416575-700 REST9416510 REST9416511 REST9416512 REST9416513 REST9416515 REST9416517 REST9416518 REST9416519 REST9416521 REST9416522 REST9416523 REST9416525 REST9416531 REST9416532 REST9416533 REST9416535 REST9416550 REST9416551 REST9416552 REST9416553 REST9416555 REST9416557 REST9416558 REST9416559 REST9416560 REST9416561 REST9416562 REST9416563 REST9416565 REST9416567 REST9416568 REST9416569 REST9416570 REST9416571 REST9416572 REST9416573 REST9416575 REST9416577 REST9416578 REST9416579 REST941650210 REST941650212 REST941650220 REST941650222 554-1561 554-1562 554-1563 REST941651E REST941652E REST941653E REST941655E REST941656E REST941657E
UHPLC columns, Pinnacle® DB Cyano (USP L10)
Chromatography Columns
Highly base-deactivated spherical silica manufactured by Restek. Cyano bonding. Suitable for analyses of a wide range of compounds, from acidic through slightly basic. Also useful for confirmation of analyses on a C18 or C8 column. Can be used in normal phase or reversed phase mode of separation.