RETSCH Test Sieve ASTM E 118x2 3.35mm 6Mesh stainless steel with calibration certificate acc. EN 10204 3.1 1 * 1 items
Supplier: Retsch GmbH
RETSCH Test Sieve ASTM E 118x2 3.35mm 6Mesh stainless steel with calibration certificate acc. EN 10204 3.1 1 * 1 items