Filter funnels, conical, with standard taper joint, VitraPOR®
Supplier: ROBU GLASFILTER
47010 47033 47011 47012 47013 47014 47020 47021 47022 47023 47024 47030 47031 47034 47032
ROBU47010EA 78.9 EUR
ROBU47010 ROBU47033 ROBU47011 ROBU47012 ROBU47013 ROBU47014 ROBU47020 ROBU47021 ROBU47022 ROBU47023 ROBU47024 ROBU47030 ROBU47031 ROBU47034 ROBU47032
Filter funnels, conical, with standard taper joint, VitraPOR®
Funnels Filtration Funnels Funnels with Conical Joints
These conical shaped funnels, made of borosilicate glass, are with standard joint core and filter disc.
  • Versatile and recyclable material
  • Resistant to most chemicals such as solvents, acids and alkalis
  • Divided into porosity classes ranging from 1,0 to 500 µm
They are ideal for sample preparation processes and filtration of small amounts.