Toluene-α-sulphonyl fluoride

Supplier: ROCHE
Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

MQ100

10837091001
ROCH10837091001EA 200 EUR
ROCH10837091001
Toluene-α-sulphonyl fluoride
Toluene-α-sulphonyl fluoride
Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Boiling Pt: 80…85 °C (0.3 mmHg)
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR