IgG1 Isotype Control (Unconjugated)

Supplier: Rockland Immunochemicals
012-001-040
ROCK012-001-040EA 400 EUR
ROCK012-001-040
IgG1 Isotype Control (Unconjugated)
Antibodies
Rat IgG1 isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: RG1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR