Anti-TYMS Sheep Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-601-199
ROCK100-601-199EA 2060 EUR
ROCK100-601-199
Anti-TYMS Sheep Polyclonal Antibody
Antibodies
This antiserum against Human Thymidylate Synthase (hTS) has been tested for use in ELISA, immunoprecipitation, immunofluorescence microscopy.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: TYMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR