Anti-Maltose Phosphorylase Goat Polyclonal Antibody
Supplier: Rockland Immunochemicals
200-101-230S
ROCK200-101-230SEA 226 EUR
ROCK200-101-230S
Anti-Maltose Phosphorylase Goat Polyclonal Antibody
Antibodies
Maltose Phosphorylase catalyzes the phosphorolysis of maltose, leading to the formation of glucose and glucose 1-P.
  • Immunogen Type: Native Protein
Anti-MALTOSE PHOSPHORYLASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum as well as purified and partially purified MaltoseMore Product Information