Anti-MAD1L1 Mouse Polyclonal Antibody (Unconjugated) [clone: 9B10]

Supplier: Rockland Immunochemicals
200-301-903
ROCK200-301-903EA 841 EUR
ROCK200-301-903
Anti-MAD1L1 Mouse Polyclonal Antibody (Unconjugated) [clone: 9B10]
Antibodies
This Protein A purified antibody has been tested for use in flow cytometry, immunoprecipitation, immunofluorescence and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a predominant band at ~ 83 kDa.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: MAD1L1
Clonality: Polyclonal
Clone: 9B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol MAD1L1
Antigen name MAD1 mitotic arrest deficient-like 1 (yeast)
Environmentally Preferable
Antigen synonyms MAD1|TXBP181|PIG9|TP53I9
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Clone 9B10
Reactivity Human
Host Mouse
Gene ID 8379
Isotype IgG2b kappa
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Immunogen This Protein A purified monoclonal antibody was produced by repeated immunizations with full-length recombinant human MAD1L1 Protein.
Format Antibody fraction
Form Liquid (sterile filtered)
Purification Protein A purified
Cross adsorption No
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Storage temperature –20 °C
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR