Anti-DNMT1 Mouse Monoclonal Antibody (Unconjugated) [clone: 60B1220.1]

Supplier: Rockland Immunochemicals
200-301-H69
ROCK200-301-H69EA 1030 EUR
ROCK200-301-H69
Anti-DNMT1 Mouse Monoclonal Antibody (Unconjugated) [clone: 60B1220.1]
Antibodies
Anti-Dnmt1 antibody is suitbale for use in ChIP, IHC (p), IP and WB. Expect a band approximately 183 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 0.1-0.5 µg/ml; Immunochemistry: 1-2 µg/ml

Type: Primary
Antigen: DNMT1
Clonality: Monoclonal
Clone: 60B1220.1
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR