Anti-MDH1 Sheep Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-601-145S
ROCK200-601-145SEA 226 EUR
ROCK200-601-145S
Anti-MDH1 Sheep Polyclonal Antibody
Antibodies
MALATE DEHYDROGENASE antibody has been assayed against 1.0 ug of Malate Dehydrogenase [Pig Heart] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG [H&L] (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100; Other Dilution: User Optimized

Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR