Anti-EIF3E Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-A62
ROCK600-401-A62EA 821 EUR
ROCK600-401-A62
Anti-EIF3E Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 48 kDa in size corresponding to eIF3S6/Int6 by western blotting.

Recommended Dilutions: ELISA: 1:80,000; Western Blot: 1:1000;

Type: Primary
Antigen: EIF3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Rat, Dog, Bovine, Monkey
Order Now

SPECIFICATIONS

Antigen symbol EIF3E
Environmentally Preferable
Antigen name Eukaryotic translation initiation factor 3, subunit E
Antigen synonyms Int6|eIF3-p46|48kDa|eIF3-p48|Eif3s6
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Monkey,Human,Bovine,Dog,Rat,Mouse
Host Rabbit
Gene ID 16341
Isotype IgG
Western blot Yes
ELISA Yes
Epitope near the C-terminus
Immunogen This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminus of mouse EIF3S6/Int6.
Format Antibody fraction
Form Liquid (sterile filtered)
Purification affinity-purified
Cross adsorption No
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Storage temperature –20 °C
Shipping temperature 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR