Anti-RPS6KA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-C66S
ROCK600-401-C66SEA 226 EUR
ROCK600-401-C66S
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:5000-1:50000; Western Blot: 1µg/ml; Other Dilution: User Optimized

Type: Primary
Antigen: RPS6KA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Horse, Bovine, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR