Anti-IgG F(ab')2 Goat Polyclonal Antibody (Peroxidase)

Supplier: Rockland Immunochemicals
610-1304
ROCK610-1304EA 384 EUR
ROCK610-1304
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Peroxidase)
Antibodies
Anti-MOUSE IgG F(ab')2 (GOAT) Antibody is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR