Anti-IgA Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
618-109-006
ROCK618-109-006EA 319 EUR
ROCK618-109-006
Anti-IgA Goat Polyclonal Antibody (Texas Red®)
Antibodies
Secondary Goat Anti-IgA Reacts with Ferret

Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Ferret
Order Now

SPECIFICATIONS

Antigen symbol IgA
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Texas Red®
Reactivity Ferret
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Immunogen Ferret IgA alpha heavy chain
Format Antibody fraction
Form Lyophilized
Purification purified by immunoaffinity chromatography
Cross adsorption none
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR