Safety shields, Sekuroka®
Supplier: Roth Carl
2967.1 2968.1
ROTH2967.1EA 0 EUR
ROTH2967.1 ROTH2968.1
Safety shields, Sekuroka®
Shields Safety Shields
These safety shields are made from transparent acrylic glass of 8 mm thick.
  • Collapsable
  • Protects against splinters and splashes
  • Three parts, can be folded out to 120°