HMDS (1,1,1,3,3,3-hexamethyldisilazane)

Supplier: Roth Carl
Danger

Synonyms: Hexamethyldisilazane

3840.1 3840.2
ROTH3840.1EA 0 EUR
ROTH3840.1 ROTH3840.2
HMDS (1,1,1,3,3,3-hexamethyldisilazane)
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 126
Melting Pt: –82 °C
Density: 0.7742 g/cm³ (20 °C)
Flash Pt: 6
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 2924
ADR: 3 II
REACH: 01-2119438176-38
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR