1,2-Dichloroethane

Supplier: Roth Carl
Danger

Synonyms: Ethylene chloride, Ethylene dichloride, Dichloroethane

6837.2 6837.3 6837.4
ROTH6837.2EA 0 EUR
ROTH6837.2 ROTH6837.3 ROTH6837.4
1,2-Dichloroethane
1,2-Dichloroethane
Formula: ClH₂CCH₂Cl
MW: 98.96 g/mol
Boiling Pt: 83.5
Melting Pt: –35.5 °C
Density: 1.254 g/cm³ (20 °C)
Flash Pt: 13
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3 II
Merck Index: 13,03831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48789e88e6041156e33712fd7c0198d7a4910266&vid=4238623c54017660b83d3baa856a69cc1aa8ee6d&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR