POPSO sesquisodium salt (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid sesquisodium salt)

Supplier: Roth Carl

Synonyms: POPSO sesquisodium, 3,3'-(Piperazine-1,4-diyl)bis(2-hydroxypropane-1-sulphonic acid), sodium salt(2:3)

6965.1 6965.2
ROTH6965.1EA 77.7 EUR
ROTH6965.1 ROTH6965.2
POPSO sesquisodium salt (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid sesquisodium salt)
POPSO sesquisodium salt (β,β'-Dihydroxypiperazine-1,4-dipropanesulphonic acid sesquisodium salt)
Formula: C₂₀H₄₁N₄Na₃O₁₆S₄
MW: 793.8099 g/mol
MDL Number: MFCD00070004
CAS Number: 108321-08-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR