Indol-3-acetic acid

Supplier: Roth Carl

Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid

7280.1
ROTH7280.1EA 24.7 EUR
ROTH7280.1
Indol-3-acetic acid
Indol-3-acetic acid
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 120…125 °C (0.05 mmHg)
Melting Pt: 167…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2
Merck Index: 13,04986

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR