Orotic acid monohydrate

Supplier: Roth Carl

Synonyms: Orotic acid hydrate, 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate

7285.1
ROTH7285.1EA 22.1 EUR
ROTH7285.1
Orotic acid monohydrate
Orotic acid monohydrate
Formula: C₅H₄N₂O₄·1H₂O
MW: 156.10 g/mol (anhy)
MDL Number: MFCD00149398
CAS Number: 50887-69-9
EINECS: 200-619-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR