Ethyl 3-phenylpropionate

Supplier: Roth Carl

Synonyms: Ethyl 3-Phenylpropionate, Ethyl hydrocinnamate, 3-Phenylpropionic acid ethyl ester, Ethyl dihydrocinnamate, Hydrocinnamic acid ethyl ester

7920.1
ROTH7920.1EA 64.2 EUR
ROTH7920.1
Ethyl 3-phenylpropionate
Ethyl 3-phenylpropionate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 247 °C (1013 hPa)
Density: 1.0343 g/cm³ (0 °C)
MDL Number: MFCD00009206
CAS Number: 2021-28-5
EINECS: 217-966-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR