Xylitol

Supplier: Roth Carl

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

9711.1 9711.2
ROTH9711.1EA 0 EUR
ROTH9711.1 ROTH9711.2
Xylitol
Xylitol
Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Boiling Pt: 495
Melting Pt: 94
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0
EINECS: 201-788-0
Merck Index: 13,10140

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR