Ammonium hydrogen carbonate

Supplier: Roth Carl
Warning

Synonyms: Ammonium bicarbonate

T871.1 T871.2 T871.5
ROTHT871.1EA 30.8 EUR
ROTHT871.1 ROTHT871.2 ROTHT871.5
Ammonium hydrogen carbonate
Ammonium hydrogen carbonate
Formula: NH₄HCO₃
MW: 79.06 g/mol
Melting Pt: 35
Density: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR