Pharmacist's spoon with wide ends
Supplier: RSG
027.200 027.150 027.280 027.320 027.240
RSGA027.200EA 42 EUR
RSGA027.200 RSGA027.150 RSGA027.280 RSGA027.320 RSGA027.240
Pharmacist's spoon with wide ends
Spoons
These pharmacist's spoons made of stainless steel (Remanit 4301) feature a broad spatula end and a spoon end.