Sodium hydroxide

Supplier: G Bioscienes
Danger

Synonyms: Caustic soda

S047
S047EA 144 EUR
S047
Sodium hydroxide
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR