Ammonium cerium(IV) nitrate

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

S33254.PG S33254.99
S33254.PGEA 0 EUR
S33254.PG S33254.99
Ammonium cerium(IV) nitrate
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 107…108 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1 III
Merck Index: 13,00509

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR