Sudan III, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: Solvent Red 23, Sudan® III (Red), Scarlet B, Sudan Red III, Cerasin Red, Sudan Red BK, Sudan Red B, Fat Ponceau G, Sudan G, D and C Red No. 17, Oil red, Tony Red, Oil scarlet, Organol® red BS, 1-(4-(Phenylazo)phenylazo)-2-naphthol, Sudan V, 1-((4-(Phenyldiazenyl)phenyl)diazenyl)naphthalen-2-ol

M-Clarity™ quality level = MQ200

S4136-25G
S4136-25GEA 148 EUR
S4136-25G
Sudan III, Sigma-Aldrich®
Sudan III
Formula: C₂₂H₁₆N₄O
MW: 352.4 g/mol
Melting Pt: 205.3…206.3 °C
MDL Number: MFCD00003905
CAS Number: 85-86-9
EINECS: 201-638-4
Merck Index: 12,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0e02492b746b52f111b5cf3a28742e424ce6d37&vid=40e67fd92d2951ca090f87fb9556557037981427&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR