Sample bags, PureBag
Supplier: Sampling Systems
8080A-450
SAMP8080A-450EA 898 EUR
SAMP8080A-450
Sample bags, PureBag
Bags Sample Bags
These are an ideal low cost alternative sterile bags for standard containers and are available in wide range of sizes.
  • Pyrogen free
  • Suitable up to 80 °C
  • Made from a special tough grade of LDPE
  • Leakproof with unique metal closure system
Certifikace: Complies with FDA, USDA and HACCP.