Sample spoons, USP Class VI, double-bagged, ULTRA
Supplier: Sampling Systems
9088H-03S 9088H-10S
SAMP9088H-03SEA 289 EUR
SAMP9088H-03S SAMP9088H-10S
Sample spoons, USP Class VI, double-bagged, ULTRA
Spoons
These sterile HDPE sampling spoons are great for taking powder and liquid samples. They are commonly used for taking sterile samples from tubs, bags, containers, bins etc.
  • Double-bagged
  • Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
The spoons are designed to be used straight from the bag. The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
Certifikace: USP Class VI Medical grade polymer is FDA and EC compliant, testedMore Product Information