Semi-micro laboratory balances, Cubis® II MCE
Supplier: Sartorius Balances
MCE225P2S00-U MCE225S2S00-U MCE125S2S00-U MCE125P2S00-U MCE225S2S00-A MCE225P2S00-A MCE125S2S00-A MCE125P2S00-A MCE225S2S00-I MCE225P2S00-I MCE125S2S00-I MCE125P2S00-I
SARBMCE225P2S00-UEA 11905 EUR
SARBMCE225P2S00-U SARBMCE225S2S00-U SARBMCE125S2S00-U SARBMCE125P2S00-U SARBMCE225S2S00-A SARBMCE225P2S00-A SARBMCE125S2S00-A SARBMCE125P2S00-A SARBMCE225S2S00-I SARBMCE225P2S00-I SARBMCE125S2S00-I SARBMCE125P2S00-I
Semi-micro laboratory balances, Cubis® II MCE
Balances and Scales
Cubis® II MCE semi-micro balances with essential user interface offer readability of 10 µg and a maximum weighing capacity up to 220 g. Used for weighing small sample amounts down to the µg-range.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent complianceMore Product Information