Microtubes
Supplier: SARSTEDT
72.694.100 72.692.105 72.730.100 72.694.106 72.694.105 72.730.106 72.693.105 72.730.105 72.692.100 72.693.100 72.694.107 72.730.107
SARS72.694.100EA 570 EUR
SARS72.694.100 SARS72.692.105 SARS72.730.100 SARS72.694.106 SARS72.694.105 SARS72.730.106 SARS72.693.105 SARS72.730.105 SARS72.692.100 SARS72.693.100 SARS72.694.107 SARS72.730.107
Microtubes
Tubes Microtubes
PP, translucent
  • Excellent chemical resistance
  • Excellent temperature resistance
Without or with skirted base.