Stripping and sealing tongs
Supplier: SARSTEDT
97.8800.200
SARS97.8800.200EA 282 EUR
SARS97.8800.200
Stripping and sealing tongs
Tongs
Stripping and sealing nippers for stripping blood bag tubes and attaching seals.